Ethyl-3-nitrobenzoat

Lieferant: CHEMPUR

Synonyme: 3-Nitrobenzoesäureethylester

BTB09265.250 BTB09265.1
CHMPBTB09265.250EA 0 EUR
CHMPBTB09265.250 MAYBBTB09265.1
Ethyl-3-nitrobenzoat
Ethyl-3-nitrobenzoat
Formel: C₉H₉NO₄
Molecular Weight: 195.17 g/mol
MDL: MFCD00014702
CAS-Nummer: 618-98-4
EINECS: 210-574-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR