Ethyl-4-ethoxybenzoat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Ethoxybenzoesäureethylester
A15010.14
A15010.22
A15010.14EA
77.8
EUR
A15010.14
A15010.22
Ethyl-4-ethoxybenzoat ≥98%
Ethyl-4-ethoxybenzoat
Purity: 98%
CAS No.: 23676-09-7
Molecular Formula: C11H14O3
Formula Weight: 194.23
Flash Point: 137°(278°F)
Refractive Index: 1.5180
Melting Point: 9°
Boiling Point: 145-150°/13mm
Beilstein Reference: 2692236
MDL No.: MFCD00009116
Formel:
C₁₁H₁₄O₃ Molecular Weight: 194,23 g/mol Siedepunkt: 145…150 °C (13 mmHg) Schmelzpunkt: 9 °C Dichte: 1,071 Flammpunkt: 137 °C (278 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009116 CAS-Nummer: 23676-09-7 EINECS: 245-818-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...