Ethylsalicylat ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Hydroxybenzoesäureethylester, Ethyl-2-hydroxybenzoat, Salicylsäureethylester

A10101.0E A10101.22 A10101.36
A10101.0EEA 149 EUR
A10101.0E A10101.22 A10101.36
Ethylsalicylat ≥99%
Ethylsalicylat

Purity: 99% CAS No.: 118-61-6 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 107°(224°F) Refractive Index: 1.5220 Melting Point: 1° Boiling Point: 232° Beilstein Reference: 907659 Merck Reference: 14,3850 MDL No.: MFCD00002215

Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Siedepunkt: 232 °C
Schmelzpunkt: 1 °C
Dichte: 1,130
Flammpunkt: 107 °C (224 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002215
CAS-Nummer: 118-61-6
EINECS: 204-265-5
Merck Index: 14,3850

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR