Dimethylphthalat 99%
Lieferant: Thermo Scientific
Synonyme:
1,2-Benzoldicarbonsäuredimethylester
, Phthalsäuredimethylester
ACRO116640025EA
111
EUR
ACRO116640025
ACRO116642500
ACRO116640250
ACRO116640010
Dimethylphthalat 99%
Dimethylphthalat
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194,19 g/mol Siedepunkt: 282 °C (760 mmHg) Dichte: 1,19 g/cm³ Flammpunkt: 150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008425 CAS-Nummer: 131-11-3 EINECS: 205-011-6 Merck Index: 13,03281 |
Spezifikation Testergebnisse
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5140 to 1.5160 (20°C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤20 APHA |
Iron (Fe) | ≤0.002 % |
Potassium (K) | ≤0.005 % |
Sodium (Na) | ≤0.02 % |
Strontium (Sr) | ≤0.7 % |
Heavy metals (as Pb) | ≤10 ppm |
Water soluble titr. base | ≤0.002 meq/g |
Insoluble matter | ≤0.015 % (in dilute HCl) |
Oxidizing substance (as NO3) | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...