Dimethylphthalat ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Phthalsäuredimethylester, 1,2-Benzoldicarbonsäuredimethylester
A14253.0E
A14253.30
A14253.36
A14253.0EEA
81.8
EUR
A14253.0E
A14253.30
A14253.36
Dimethylphthalat ≥99%
Dimethylphthalat
Purity: 99%
CAS No.: 131-11-3
Molecular Formula: C10H10O4
Formula Weight: 194.19
Flash Point: 146°(294°F)
Physical Form: Liquid
Density: 1.190
Refractive Index: 1.5150
Melting Point: ca 5°
Boiling Point: 283-284°
Beilstein Reference: 1911460
Merck Reference: 14,3255
MDL No.: MFCD00008425
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194.19 g/mol Siedepunkt: 283…284 °C Schmelzpunkt: ∼ 5 °C Dichte: 1.190 Flammpunkt: 146 °C (294 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008425 CAS-Nummer: 131-11-3 EINECS: 205-011-6 Merck Index: 13,03281 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...