Kaliumbenzoat ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: Benzoic acid potassium salt

A11610.0B A11610.30 AB118625.0250
A11610.0BEA 110 EUR
A11610.0B A11610.30 ABCRAB118625.0250
Kaliumbenzoat ≥99%
Kaliumbenzoat

Purity: 99% CAS No.: 582-25-2 Molecular Formula: C7H5KO2 Formula Weight: 160.22 Physical Form: Powder Sensitivity: Hygroscopic Beilstein Reference: 3719165 Merck Reference: 14,1091 MDL No.: MFCD00013061

Formel: C₇H₅KO₂
Molecular Weight: 160.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013061
CAS-Nummer: 582-25-2
EINECS: 209-481-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR