5-Methoxyanthranilsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
L10295.03
L10295.06
L10295.03EA
75.3
EUR
L10295.03
L10295.06
5-Methoxyanthranilsäure ≥98%
5-Methoxyanthranilsäure
Purity: 98+%
CAS No.: 6705-03-9
Molecular Formula: C8H9NO3
Formula Weight: 167.16
Melting Point: 150-152°
Beilstein Reference: 777690
Formel:
C₈H₉NO₃ Molecular Weight: 167,16 g/mol Schmelzpunkt: 150…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016509 CAS-Nummer: 6705-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...