5-Methoxyanthranilsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

665118-1G 665118-5G
SIAL665118-1GEA 93.4 EUR
SIAL665118-1G SIAL665118-5G
5-Methoxyanthranilsäure, Sigma-Aldrich®
5-Methoxyanthranilsäure
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016509
CAS-Nummer: 6705-03-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR