6-Methoxyanthranilsäure

Lieferant: FLUOROCHEM

009747-1G 009747-5G
FLUO009747-1GEA 36.9 EUR
FLUO009747-1G FLUO009747-5G
6-Methoxyanthranilsäure
6-Methoxyanthranilsäure
Formel: C₈H₉NO₃
Molecular Weight: 167.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01941328
CAS-Nummer: 53600-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR