4-Amino-o-anissäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Amino-2-methoxybenzenecarboxylic acid
H27419.06
H27419.14
H27419.06EA
24.4
EUR
H27419.06
H27419.14
4-Amino-o-anissäure 97%
4-Amino-o-anissäure
Purity: 97%
CAS No.: 2486-80-8
Molecular Formula: C8H9NO3
Formula Weight: 167.16
Melting Point: ca 150° dec.
MDL No.: MFCD00114479
Formel:
C₈H₉NO₃ Molecular Weight: 167,16 g/mol Schmelzpunkt: ∼ 150 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00114479 CAS-Nummer: 2486-80-8 EINECS: 219-632-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...