4-Amino-o-anissäure

Lieferant: Thermo Scientific

Synonyme: 4-Amino-2-methoxybenzenecarboxylic acid

XBX00151.250 XBX00151.1
MAYBXBX00151.250EA 0 EUR
MAYBXBX00151.250 MAYBXBX00151.1
4-Amino-o-anissäure
4-Amino-o-anissäure
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
MDL: MFCD00114479
CAS-Nummer: 2486-80-8
EINECS: 219-632-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR