4-Amino-o-anissäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-2-methoxybenzenecarboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

647624-25G 647624-5G
SIAL647624-25GEA 58.2 EUR
SIAL647624-25G SIAL647624-5G
4-Amino-o-anissäure, Sigma-Aldrich®
4-Amino-o-anissäure
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
MDL: MFCD00114479
CAS-Nummer: 2486-80-8
EINECS: 219-632-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR