3,4,5-Trimethoxyanthranilsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

631272-25G 631272-5G
SIAL631272-25GEA 350 EUR
SIAL631272-25G SIAL631272-5G
3,4,5-Trimethoxyanthranilsäure, Sigma-Aldrich®
3,4,5-Trimethoxyanthranilsäure
Formel: C₁₀H₁₃NO₅
Molecular Weight: 227,22 g/mol
MDL: MFCD00051630
CAS-Nummer: 61948-85-4
EINECS: 263-344-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR