N-Phenylanthranilsäure ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(Phenylamino)benzoesäure
A14340.22
A14340.14
A14340.36
A14340.22EA
97.9
EUR
A14340.22
A14340.14
A14340.36
N-Phenylanthranilsäure ≥99%
N-Phenylanthranilsäure
Purity: 99%
CAS No.: 91-40-7
Molecular Formula: C13H11NO2
Formula Weight: 213.24
Melting Point: 185-188°
Beilstein Reference: 1456607
Merck Reference: 14,7273
MDL No.: MFCD00002421
Formel:
C₁₃H₁₁NO₂ Molecular Weight: 213.24 g/mol Siedepunkt: 375 Schmelzpunkt: 185…188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002421 CAS-Nummer: 91-40-7 EINECS: 202-066-8 Merck Index: 12,07427 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...