4,5-Dimethoxy-2-nitrobenzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L08510.14 L08510.22 L08510.06
L08510.14EA 68 EUR
L08510.14 L08510.22 L08510.06
4,5-Dimethoxy-2-nitrobenzoesäure ≥98%
4,5-Dimethoxy-2-nitrobenzoesäure

Purity: 98% CAS No.: 4998-07-6 Molecular Formula: C9H9NO6 Formula Weight: 227.17 Melting Point: 192-195° Beilstein Reference: 2123903

Formel: C₉H₉NO₆
Molecular Weight: 227.17 g/mol
Schmelzpunkt: 192…195 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014697
CAS-Nummer: 4998-07-6
EINECS: 225-657-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR