3-Ethoxy-4-methoxybenzoesäure

Lieferant: FLUOROCHEM

014650-1G 014650-5G
FLUO014650-1GEA 1130 EUR
FLUO014650-1G FLUO014650-5G
3-Ethoxy-4-methoxybenzoesäure
3-Ethoxy-4-methoxybenzoesäure
Formel: C₁₀H₁₂O₃
Molecular Weight: 180.2 g/mol
MDL: MFCD00276591
CAS-Nummer: 2651-55-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR