2-(Methylthio)benzoesäure
Lieferant: FLUOROCHEM
Synonyme:
2-(Methylmercapto)benzoesäure
002628-1G
002628-5G
FLUO002628-1GEA
31.1
EUR
FLUO002628-1G
FLUO002628-5G
2-(Methylthio)benzoesäure
2-(Methylthio)benzoesäure
Formel:
C₈H₈O₂S Molecular Weight: 168,22 g/mol |
MDL:
MFCD00029934 CAS-Nummer: 3724-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...