Kaliumhydrogenphthalat 99,95-100,05% ACS primärer Standard
Lieferant: Thermo Scientific
Achtung
Synonyme: Phthalsäure Kaliumsalz, 1,2-Benzoldicarbonsäure Kaliumsalz, Kaliumphthalat monobasisch
33351.30 33351.A1
33351.30EA 68.5 EUR
33351.30 33351.A1
Kaliumhydrogenphthalat 99,95-100,05% ACS primärer Standard
Kaliumhydrogenphthalat
Purity: 99.95-100.05 CAS No.: 877-24-7 Molecular Formula: C8H5KO4 Formula Weight: 204.23 Physical Form: Crystalline powder Density: 1.636 Appearance: White Odour: Odourless Melting Point: 297° dec. Beilstein Reference: 3637128 Merck Reference: 14,7612 MDL No.: MFCD00013070
As a primary standard for preparing volumetric alkali solutions and as a buffer in pH determinations
Bitte beachten Incompatible with strong oxidising agents.
Formel: HOOCC₆H₄COOK
Molecular Weight: 204,22 g/mol
Schmelzpunkt: 297 °C (Zers.)
Dichte: 1,636
Lagertemperatur: Raumtemperatur
MDL: MFCD00013070
CAS-Nummer: 877-24-7
EINECS: 212-889-4
Merck Index: 14,7612
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=75071da7f30b07f7bd077699a3b54ec89d80f0b8&prid=web&clid=SAPDAM