2,3-Difluorphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: Dihydroxy(2,3-difluorophenyl)borane, 2,3-Difluorobenzeneboronic acid, (2,3-Difluorophenyl)metaboric acid

013811-5G 013811-25G 13811.25
FLUO013811-5GEA 45.9 EUR
FLUO013811-5G FLUO013811-25G CHMP13811.25
2,3-Difluorphenylboronsäure
2,3-Difluorphenylboronsäure
Formel: C₆H₅BF₂O₂
Molecular Weight: 157.91 g/mol
MDL: MFCD01863170
CAS-Nummer: 121219-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR