2,4,5-Trifluorphenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27858.14
H27858.06
H27858.14EA
504
EUR
H27858.14
H27858.06
2,4,5-Trifluorphenylboronsäure ≥97%
2,4,5-Trifluorphenylboronsäure
Purity: 97%
CAS No.: 247564-72-3
Molecular Formula: C6H4BF3O2
Formula Weight: 175.90
Melting Point: 99-101°
MDL No.: MFCD01863165
Formel:
C₆H₄BF₃O₂ Molecular Weight: 175.9 g/mol Schmelzpunkt: 99…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863165 CAS-Nummer: 247564-72-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...