2,4,6-Trifluorphenylboronsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(2,4,6-Trifluorophenyl)metaboric acid, 2,4,6-Trifluorobenzeneboronic acid, Dihydroxy(2,4,6-trifluorophenyl)borane
L19402.03
L19402.06
L19402.03EA
35.8
EUR
L19402.03
L19402.06
2,4,6-Trifluorphenylboronsäure ≥97%
2,4,6-Trifluorphenylboronsäure
Purity: 97%
CAS No.: 182482-25-3
Molecular Formula: C6H4BF3O2
Formula Weight: 175.90
Melting Point: ca 235°
Beilstein Reference: 8549766
Formel:
C₆H₄BF₃O₂ Molecular Weight: 175,9 g/mol Schmelzpunkt: ∼ 235 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863169 CAS-Nummer: 182482-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...