2,4,6-Trifluorphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2,4,6-Trifluorophenyl)metaboric acid, 2,4,6-Trifluorobenzeneboronic acid, Dihydroxy(2,4,6-trifluorophenyl)borane

M-Clarity™ Qualitätsstufe = MQ100

524107-1G 524107-5G
SIAL524107-1GEA 25.4 EUR
SIAL524107-1G SIAL524107-5G
2,4,6-Trifluorphenylboronsäure, Sigma-Aldrich®
2,4,6-Trifluorphenylboronsäure
Formel: C₆H₄BF₃O₂
Molecular Weight: 175,9 g/mol
MDL: MFCD01863169
CAS-Nummer: 182482-25-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR