2,4-Difluorphenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B23821.06 B23821.14
B23821.06EA 68.4 EUR
B23821.06 B23821.14
2,4-Difluorphenylboronsäure ≥97%
2,4-Difluorphenylboronsäure

Purity: 97% CAS No.: 144025-03-6 Molecular Formula: C6H5BF2O2 Formula Weight: 157.91 Melting Point: 247-250° Beilstein Reference: 8616257

Formel: C₆H₅BF₂O₂
Molecular Weight: 157.91 g/mol
Schmelzpunkt: 247…250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318998
CAS-Nummer: 144025-03-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR