2,5-Difluorphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Difluorbenzolboronsäure, (2,5-Difluorphenyl)metaborsäure, Dihydroxy(2,5-difluorphenyl)boran

M-Clarity™ Qualitätsstufe = MQ100

514020-25G 514020-5G
SIAL514020-25GEA 0 EUR
SIAL514020-25G SIAL514020-5G
2,5-Difluorphenylboronsäure, Sigma-Aldrich®
2,5-Difluorphenylboronsäure
Formel: C₆H₅BF₂O₂
Molecular Weight: 157,91 g/mol
MDL: MFCD01863171
CAS-Nummer: 193353-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR