2,6-Difluorphenylboronsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: Dihydroxy(2,6-difluorophenyl)borane, 2,6-Difluorobenzeneboronic acid, (2,6-Difluorophenyl)metaboric acid

005228-1G 005228-5G
FLUO005228-1GEA 30.8 EUR
FLUO005228-1G FLUO005228-5G
2,6-Difluorphenylboronsäure
2,6-Difluorphenylboronsäure
Formel: C₆H₅BF₂O₂
Molecular Weight: 157.91 g/mol
MDL: MFCD00792436
CAS-Nummer: 162101-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR