2-Fluorphenylboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(2-Fluorphenyl)metaborsäure, 2-Fluorbenzolboronsäure, Dihydroxy(2-fluorphenyl)boran
B23103.03
B23103.06
B23103.14
B23103.03EA
48.4
EUR
B23103.03
B23103.06
B23103.14
2-Fluorphenylboronsäure ≥98%
2-Fluorphenylboronsäure
Purity: 98%
CAS No.: 1993-03-9
Molecular Formula: C6H6BFO2
Formula Weight: 139.92
Melting Point: 100-108°
Beilstein Reference: 3030413
Formel:
C₆H₆BFO₂ Molecular Weight: 139.92 g/mol Schmelzpunkt: 100…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00674013 CAS-Nummer: 1993-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...