3,4,5-Trifluorphenylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (3,4,5-Trifluorphenyl)metaborsäure, 3,4,5-Trifluorbenzolboronsäure, Dihydroxy(3,4,5-trifluorphenyl)boran

L18519.03 L18519.MD
L18519.03EA 160 EUR
L18519.03 L18519.MD
3,4,5-Trifluorphenylboronsäure ≥97%
3,4,5-Trifluorphenylboronsäure

Purity: 97% CAS No.: 143418-49-9 Molecular Formula: C6H4BF3O2 Formula Weight: 175.90 Melting Point: 275-279° Beilstein Reference: 7371914

Formel: C₆H₄BF₃O₂
Molecular Weight: 175,9 g/mol
Schmelzpunkt: 275…279 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093069
CAS-Nummer: 143418-49-9
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR