3,4,5-Trifluorphenylboronsäure
Lieferant: FLUOROCHEM
Synonyme:
(3,4,5-Trifluorphenyl)metaborsäure, 3,4,5-Trifluorbenzolboronsäure, Dihydroxy(3,4,5-trifluorphenyl)boran
013045-5G
013045-25G
013045-50G
FLUO013045-5GEA
45.9
EUR
FLUO013045-5G
FLUO013045-25G
FLUO013045-50G
3,4,5-Trifluorphenylboronsäure
3,4,5-Trifluorphenylboronsäure
Formel:
C₆H₄BF₃O₂ Molecular Weight: 175,9 g/mol Schmelzpunkt: 292 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093069 CAS-Nummer: 143418-49-9 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...