3,4-Difluorphenylboronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,4-Difluorbenzolboronsäure, Dihydroxy(3,4-difluorphenyl)boran, (3,4-Difluorphenyl)metaborsäure
AC35916.1
AC35916.50
359160050
MAYBAC35916.1EA
0
EUR
MAYBAC35916.1
MAYBAC35916.50
ACRO359160050
3,4-Difluorphenylboronsäure 97%
3,4-Difluorphenylboronsäure
Formel:
C₆H₅BF₂O₂ Molecular Weight: 157,91 g/mol Schmelzpunkt: 289…290 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00807405 CAS-Nummer: 168267-41-2 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...