3,4-Difluorphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 3,4-Difluorbenzolboronsäure, Dihydroxy(3,4-difluorphenyl)boran, (3,4-Difluorphenyl)metaborsäure

013040-25G 013040-5G
FLUO013040-25GEA 63.9 EUR
FLUO013040-25G FLUO013040-5G
3,4-Difluorphenylboronsäure
3,4-Difluorphenylboronsäure
Formel: C₆H₅BF₂O₂
Molecular Weight: 157.91 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00807405
CAS-Nummer: 168267-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR