3,5-Difluorphenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (3,5-Difluorophenyl)metaboric acid, Dihydroxy(3,5-difluorophenyl)borane, 3,5-Difluorobenzeneboronic acid

L17425.03 L17425.06
L17425.03EA 30.7 EUR
L17425.03 L17425.06
3,5-Difluorphenylboronsäure ≥97%
3,5-Difluorphenylboronsäure

Purity: 97+% CAS No.: 156545-07-2 Molecular Formula: C6H5BF2O2 Formula Weight: 157.91 Melting Point: 300° Beilstein Reference: 6800882

Formel: C₆H₅BF₂O₂
Molecular Weight: 157.91 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01318138
CAS-Nummer: 156545-07-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR