3-Fluorphenylboronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(3-Fluorphenyl)metaborsäure, 3-Fluorbenzolboronsäure, Dihydroxy(3-fluorphenyl)boran
345640010
345640050
AC34564.1
AC34564.10
345640250
ACRO345640010EA
30.7
EUR
ACRO345640010
ACRO345640050
MAYBAC34564.1
MAYBAC34564.10
ACRO345640250
3-Fluorphenylboronsäure 97%
3-Fluorphenylboronsäure
Formel:
C₆H₆BFO₂ Molecular Weight: 139,92 g/mol Schmelzpunkt: 211…212 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00236042 CAS-Nummer: 768-35-4 |
Spezifikation Testergebnisse
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...