4-Fluorphenylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Dihydroxy(4-fluorphenyl)boran, 4-Fluorbenzolboronsäure, (4-Fluorphenyl)metaborsäure

A15991.03 A15991.06 A15991.14
A15991.03EA 17.8 EUR
A15991.03 A15991.06 A15991.14
4-Fluorphenylboronsäure ≥97%
4-Fluorphenylboronsäure

Purity: 97% CAS No.: 1765-93-1 Molecular Formula: C6H6BFO2 Formula Weight: 139.92 Melting Point: 260-265° Beilstein Reference: 2829653

Formel: C₆H₆BFO₂
Molecular Weight: 139,92 g/mol
Schmelzpunkt: 260…265 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039136
CAS-Nummer: 1765-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR