4-Fluorphenylboronsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: Dihydroxy(4-fluorphenyl)boran, 4-Fluorbenzolboronsäure, (4-Fluorphenyl)metaborsäure

006182-5G 006182-25G 006182-100G
FLUO006182-5GEA 0 EUR
FLUO006182-5G FLUO006182-25G FLUO006182-100G
4-Fluorphenylboronsäure
4-Fluorphenylboronsäure
Formel: C₆H₆BFO₂
Molecular Weight: 139.92 g/mol
Schmelzpunkt: 263…265 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039136
CAS-Nummer: 1765-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR