2-(Trifluormethoxy)phenylboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(2-(Trifluormethoxy)phenyl)metaborsäure, 2-(Trifluormethoxy)benzolboronsäure, Dihydroxy(2-(trifluormethoxy)phenyl)boran
M-Clarity™ Qualitätsstufe = MQ100
683787-1G
683787-5G
SIAL683787-1GEA
29.2
EUR
SIAL683787-1G
SIAL683787-5G
2-(Trifluormethoxy)phenylboronsäure, Sigma-Aldrich®
2-(Trifluormethoxy)phenylboronsäure
Formel:
C₇H₆BF₃O₃ Molecular Weight: 205.93 g/mol |
MDL:
MFCD01320763 CAS-Nummer: 175676-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...