4-(Trifluormethoxy)phenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: Dihydroxy(4-(trifluormethoxy)phenyl)boran, 4-(Trifluormethoxy)benzolboronsäure, (4-(Trifluormethoxy)phenyl)metaborsäure

013041-25G 013041-5G
FLUO013041-25GEA 159 EUR
FLUO013041-25G FLUO013041-5G
4-(Trifluormethoxy)phenylboronsäure
4-(Trifluormethoxy)phenylboronsäure
Formel: C₇H₆BF₃O₃
Molecular Weight: 205.93 g/mol
MDL: MFCD01074648
CAS-Nummer: 139301-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR