2,4,6-Trifluorbenzaldehyd

Lieferant: FLUOROCHEM
Achtung

013013-1G 013013-5G 013013-25G
FLUO013013-1GEA 30.8 EUR
FLUO013013-1G FLUO013013-5G FLUO013013-25G
2,4,6-Trifluorbenzaldehyd
2,4,6-Trifluorobenzaldehyde
Formel: C₇H₃F₃O
Molecular Weight: 160.1 g/mol
MDL: MFCD00061197
CAS-Nummer: 58551-83-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR