2-(Difluormethoxy)benzaldehyd

Lieferant: FLUOROCHEM
Achtung

004829-5G 004829-25G
FLUO004829-5GEA 0 EUR
FLUO004829-5G FLUO004829-25G
2-(Difluormethoxy)benzaldehyd
2-(Difluormethoxy)benzaldehyd
Formel: C₈H₆F₂O₂
Molecular Weight: 172,13 g/mol
MDL: MFCD00042251
CAS-Nummer: 71653-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR