2-(Trifluormethoxy)benzaldehyd 96%

Lieferant: Thermo Scientific
Achtung

B20840.06 B20840.14 B20840.22
B20840.06EA 49.7 EUR
B20840.06 B20840.14 B20840.22
2-(Trifluormethoxy)benzaldehyd 96%
2-(Trifluormethoxy)benzaldehyd

Purity: 96% CAS No.: 94651-33-9 Molecular Formula: C8H5F3O2 Formula Weight: 190.12 Flash Point: 67°(152°F) Refractive Index: 1.4550 Boiling Point: 77°/20mm Beilstein Reference: 6137162 MDL No.: MFCD00042405

Formel: C₈H₅F₃O₂
Molecular Weight: 190,12 g/mol
Siedepunkt: 77 °C (20 mmHg)
Dichte: 1,330
Flammpunkt: 67 °C (152 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042405
CAS-Nummer: 94651-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR