3-(Difluormethoxy)benzaldehyd

Lieferant: FLUOROCHEM
Achtung

007851-1G 007851-5G
FLUO007851-1GEA 51.8 EUR
FLUO007851-1G FLUO007851-5G
3-(Difluormethoxy)benzaldehyd
3-(Difluormethoxy)benzaldehyd
Formel: C₈H₆F₂O₂
Molecular Weight: 172,13 g/mol
MDL: MFCD00236222
CAS-Nummer: 85684-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR