3-Fluor-p-anisaldehyd, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Fluor-4-methoxybenzaldehyd, 3-Fluor-4-anisaldehyd
M-Clarity™ Qualitätsstufe = MQ100
155586-1G
155586-5G
SIAL155586-1GEA
58.6
EUR
SIAL155586-1G
SIAL155586-5G
3-Fluor-p-anisaldehyd, Sigma-Aldrich®
3-Fluor-p-anisaldehyd
Formel:
C₈H₇FO₂ Molecular Weight: 154.14 g/mol |
MDL:
MFCD00003349 CAS-Nummer: 351-54-2 EINECS: 206-514-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...