2,3,4,5-Tetrafluorbenzonitril

Lieferant: FLUOROCHEM

005843-1G 005843-5G
FLUO005843-1GEA 34.1 EUR
FLUO005843-1G FLUO005843-5G
2,3,4,5-Tetrafluorbenzonitril
2,3,4,5-Tetrafluorbenzonitril
Formel: C₇HF₄N
Molecular Weight: 175,09 g/mol
Siedepunkt: 164…165 °C (1013 hPa)
Dichte: 1,463 g/cm³ (20 °C)
MDL: MFCD00040187
CAS-Nummer: 16582-93-7
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR