2,3,4-Trifluorbenzonitril

Lieferant: FLUOROCHEM

008712-5G 008712-25G 008712-100G
FLUO008712-5GEA 0 EUR
FLUO008712-5G FLUO008712-25G FLUO008712-100G
2,3,4-Trifluorbenzonitril
2,3,4-Trifluorbenzonitril
Formel: C₇H₂F₃N
Molecular Weight: 157.09 g/mol
Siedepunkt: 116…118 °C (100 mmHg)
Dichte: 1.12 g/cm³ (20 °C)
MDL: MFCD00013288
CAS-Nummer: 143879-80-5
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR