2,3,6-Trifluorbenzonitril

Lieferant: FLUOROCHEM

007486-1G 007486-5G
FLUO007486-1GEA 31.1 EUR
FLUO007486-1G FLUO007486-5G
2,3,6-Trifluorbenzonitril
2,3,6-Trifluorbenzonitril
Formel: C₇H₂F₃N
Molecular Weight: 157.09 g/mol
Siedepunkt: 179
Dichte: 1.359 g/cm³ (20 °C)
MDL: MFCD00012287
CAS-Nummer: 136514-17-5
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR