3,4,5-Trifluorbenzonitril

Lieferant: FLUOROCHEM

007488-5G 007488-1G
FLUO007488-5GEA 0 EUR
FLUO007488-5G FLUO007488-1G
3,4,5-Trifluorbenzonitril
3,4,5-Trifluorbenzonitril
Formel: C₇H₂F₃N
Molecular Weight: 157,09 g/mol
Siedepunkt: 72…73 °C (10 mmHg)
Schmelzpunkt: 45…47 °C
MDL: MFCD00074948
CAS-Nummer: 134227-45-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR