2-(Trifluormethoxy)benzonitril

Lieferant: CHEMPUR

FL7544.1 007544-5G 007544-25G 007544-100G
CHMPFL7544.1EA 0 EUR
CHMPFL7544.1 FLUO007544-5G FLUO007544-25G FLUO007544-100G
2-(Trifluormethoxy)benzonitril
2-(Trifluormethoxy)benzonitril
Formel: C₈H₄F₃NO
Molecular Weight: 187.12 g/mol
Siedepunkt: 80…82 °C (15 mmHg)
MDL: MFCD00042409
CAS-Nummer: 63968-85-4
EINECS: 264-584-0
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR