3-(Trifluormethoxy)benzonitril
Lieferant: FLUOROCHEM
001489-1G
001489-5G
001489-25G
FLUO001489-1GEA
31.1
EUR
FLUO001489-1G
FLUO001489-5G
FLUO001489-25G
3-(Trifluormethoxy)benzonitril
3-(Trifluormethoxy)benzonitril
Formel:
C₈H₄F₃NO Molecular Weight: 187.12 g/mol Siedepunkt: 236 Schmelzpunkt: 37 |
MDL:
MFCD00040959 CAS-Nummer: 52771-22-9 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...