2,3,5-Trifluorbenzoylchlorid

Lieferant: FLUOROCHEM

007960-5G 007960-1G
FLUO007960-5GEA 296 EUR
FLUO007960-5G FLUO007960-1G
2,3,5-Trifluorbenzoylchlorid
2,3,5-Trifluorbenzoylchlorid
Formel: C₇H₂ClF₃O
Molecular Weight: 194.54 g/mol
CAS-Nummer: 240800-48-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR