2,4,5-Trifluorbenzoylchlorid
Lieferant: CHEMPUR
|
Gefahr
|
Synonyme:
2,4,5-Trifluorbenzoesäurechlorid
SB02059.1
SB02059.10
SB02059.50
CHMPSB02059.1EA
0
EUR
CHMPSB02059.1
CHMPSB02059.10
CHMPSB02059.50
2,4,5-Trifluorbenzoylchlorid
2,4,5-Trifluorbenzoylchlorid
Formel:
C₇H₂ClF₃O Molecular Weight: 194,54 g/mol Siedepunkt: 85 °C (17 mmHg) Dichte: 1,52 g/cm³ (20 °C) |
MDL:
MFCD00061204 CAS-Nummer: 88419-56-1 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...