2,4,6-Trifluorbenzoylchlorid ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2,4,6-Trifluorbenzoesäurechlorid
B20323.03
B20323.06
B20323.03EA
47.9
EUR
B20323.03
B20323.06
2,4,6-Trifluorbenzoylchlorid ≥97%
2,4,6-Trifluorbenzoylchlorid
Purity: 97%
CAS No.: 79538-29-7
Molecular Formula: C7H2ClF3O
Formula Weight: 194.54
Refractive Index: 1.4840
Boiling Point: 167-168°
UN No.: 1760
MDL No.: MFCD00070810
Formel:
C₇H₂ClF₃O Molecular Weight: 194.54 g/mol Siedepunkt: 167…168 °C Schmelzpunkt: 41 Dichte: 1.499 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070810 CAS-Nummer: 79538-29-7 UN: 1760 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...